CH4424852133 ChemBridge Corporation

2-[2-(dimethylamino)ethoxy]benzaldehyde hydrochloride

  • MDL number MFCD26959589
  • Cas Number : 15182-07-7
  • Molecular Weight : 229.7032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070947

Safety Information

MSDS https://www.hit2lead.com/sds/9070947_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart