CH4424852140 ChemBridge Corporation

N-[4-(methylthio)benzyl]cyclopropanamine hydrochloride

  • MDL number MFCD13194092
  • Cas Number : 1049696-51-6
  • Molecular Weight : 229.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070983

Safety Information

MSDS https://www.hit2lead.com/sds/9070983_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart