CH4424852143 ChemBridge Corporation

3-[(4-ethyl-1-piperazinyl)methyl]-2,5-dimethylbenzaldehyde dihydrochloride

  • MDL number MFCD13186627
  • Cas Number : 1609406-67-8
  • Molecular Weight : 333.2964

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070991

Safety Information

MSDS https://www.hit2lead.com/sds/9070991_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart