CH4424852160 ChemBridge Corporation

(2-ethoxy-3-methoxybenzyl)methylamine hydrochloride

  • MDL number MFCD07108391
  • Cas Number : 1048947-99-4
  • Molecular Weight : 231.7191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071069

Safety Information

MSDS https://www.hit2lead.com/sds/9071069_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart