CH4424852162 ChemBridge Corporation

N-(2-chlorobenzyl)-2-methyl-1-propanamine hydrochloride

  • MDL number MFCD07109823
  • Cas Number : 1050076-04-4
  • Molecular Weight : 234.1654

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071072

Safety Information

MSDS https://www.hit2lead.com/sds/9071072_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart