CH4424852173 ChemBridge Corporation

2-[(2-fluorobenzyl)amino]-2-methyl-1-propanol hydrochloride

  • MDL number MFCD07109987
  • Cas Number : 1050076-28-2
  • Molecular Weight : 233.7102

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071152

Safety Information

MSDS https://www.hit2lead.com/sds/9071152_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart