CH4424852177 ChemBridge Corporation

N-[(5-methyl-2-thienyl)methyl]cyclopropanamine hydrochloride

  • MDL number MFCD07111365
  • Cas Number : 1050214-04-4
  • Molecular Weight : 203.7322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071193

Safety Information

MSDS https://www.hit2lead.com/sds/9071193_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart