CH4424852178 ChemBridge Corporation

[2-(mesityloxy)ethyl]amine hydrochloride

  • MDL number MFCD06149949
  • Cas Number : 1048664-12-5
  • Molecular Weight : 215.7197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071196

Safety Information

MSDS https://www.hit2lead.com/sds/9071196_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart