CH4424852181 ChemBridge Corporation

tert-butyl(4-pyridinylmethyl)amine hydrochloride

  • MDL number MFCD11841391
  • Cas Number : 1185150-94-0
  • Molecular Weight : 200.7084

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071212

Safety Information

MSDS https://www.hit2lead.com/sds/9071212_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart