CH4424852187 ChemBridge Corporation

N-(3-bromobenzyl)-2-butanamine hydrochloride

  • MDL number MFCD07107537
  • Cas Number : 1049678-18-3
  • Molecular Weight : 278.6164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071250

Safety Information

MSDS https://www.hit2lead.com/sds/9071250_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart