CH4424852194 ChemBridge Corporation

N-(2-methoxybenzyl)-2-propen-1-amine hydrochloride

  • MDL number MFCD07107846
  • Cas Number : 1049678-37-6
  • Molecular Weight : 213.7038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071283

Safety Information

MSDS https://www.hit2lead.com/sds/9071283_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart