CH4424852200 ChemBridge Corporation

4-(3-methoxyphenyl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD26959444
  • Cas Number : 93690-16-5
  • Molecular Weight : 225.6748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071301

Safety Information

MSDS https://www.hit2lead.com/sds/9071301_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart