CH4424852209 ChemBridge Corporation

N-(3-phenyl-2-propyn-1-yl)cyclopropanamine hydrochloride

  • MDL number MFCD07112058
  • Cas Number : 1050126-53-8
  • Molecular Weight : 207.6993

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071364

Safety Information

MSDS https://www.hit2lead.com/sds/9071364_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart