CH4424852211 ChemBridge Corporation

N,N-dimethyl-N'-[(5-methyl-2-thienyl)methyl]-1,2-ethanediamine dihydrochloride

  • MDL number MFCD07111363
  • Cas Number : 1050213-96-1
  • Molecular Weight : 271.2502

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071368

Safety Information

MSDS https://www.hit2lead.com/sds/9071368_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart