CH4424852217 ChemBridge Corporation

[3-(2,6-dimethylphenoxy)propyl]methylamine hydrochloride

  • MDL number MFCD26959501
  • Cas Number : 1609409-05-3
  • Molecular Weight : 229.7463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071460

Safety Information

MSDS https://www.hit2lead.com/sds/9071460_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart