CH4424852221 ChemBridge Corporation

N,N-dimethyl-N'-(4-methylbenzyl)-1,2-ethanediamine dihydrochloride

  • MDL number MFCD07110790
  • Cas Number : 1158367-04-4
  • Molecular Weight : 265.2225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071479

Safety Information

MSDS https://www.hit2lead.com/sds/9071479_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart