CH4424852228 ChemBridge Corporation

N-(3-methoxybenzyl)-2-butanamine hydrochloride

  • MDL number MFCD07108534
  • Cas Number : 1051363-51-9
  • Molecular Weight : 229.7463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071525

Safety Information

MSDS https://www.hit2lead.com/sds/9071525_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart