CH4424852234 ChemBridge Corporation

N-[(3-methyl-2-thienyl)methyl]-1-butanamine hydrochloride

  • MDL number MFCD07105470
  • Cas Number : 1048664-69-2
  • Molecular Weight : 219.7746

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071564

Safety Information

MSDS https://www.hit2lead.com/sds/9071564_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart