CH4424852250 ChemBridge Corporation

(2E)-N-(2-methoxyethyl)-3-phenyl-2-propen-1-amine hydrochloride

  • MDL number MFCD07111217
  • Cas Number : 1159700-18-1
  • Molecular Weight : 227.7304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071662

Safety Information

MSDS https://www.hit2lead.com/sds/9071662_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart