CH4424852252 ChemBridge Corporation

N,N-dimethyl-N'-(3-thienylmethyl)-1,2-ethanediamine dihydrochloride

  • MDL number MFCD07170382
  • Cas Number : 1049713-58-7
  • Molecular Weight : 257.2236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071672

Safety Information

MSDS https://www.hit2lead.com/sds/9071672_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart