CH4424852260 ChemBridge Corporation

4-[(diethylamino)methyl]-5-ethyl-2-furoic acid hydrochloride

  • MDL number MFCD11506487
  • Cas Number : 1052532-17-8
  • Molecular Weight : 261.7451

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071724

Safety Information

MSDS https://www.hit2lead.com/sds/9071724_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart