CH4424852268 ChemBridge Corporation

N-(3-thienylmethyl)-1-propanamine hydrochloride

  • MDL number MFCD07170359
  • Cas Number : 1049713-35-0
  • Molecular Weight : 191.7215

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071826

Safety Information

MSDS https://www.hit2lead.com/sds/9071826_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart