CH4424852272 ChemBridge Corporation

3-{[(5-methyl-2-thienyl)methyl]amino}-1-propanol hydrochloride

  • MDL number MFCD08596491
  • Cas Number : 1050502-69-6
  • Molecular Weight : 221.7474

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071843

Safety Information

MSDS https://www.hit2lead.com/sds/9071843_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart