CH4424852273 ChemBridge Corporation

(2-methoxyethyl)(3-pyridinylmethyl)amine hydrochloride

  • MDL number MFCD07110860
  • Cas Number : 1049803-09-9
  • Molecular Weight : 202.6812

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071847

Safety Information

MSDS https://www.hit2lead.com/sds/9071847_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart