CH4424852275 ChemBridge Corporation

2,5-dimethyl-3-[(4-methyl-1-piperazinyl)methyl]benzaldehyde dihydrochloride

  • MDL number MFCD13186639
  • Cas Number : 1349716-60-4
  • Molecular Weight : 319.2699

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071858

Safety Information

MSDS https://www.hit2lead.com/sds/9071858_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart