CH4424852283 ChemBridge Corporation

N-(3-thienylmethyl)-2-butanamine hydrochloride

  • MDL number MFCD07170364
  • Cas Number : 1049713-37-2
  • Molecular Weight : 205.748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071896

Safety Information

MSDS https://www.hit2lead.com/sds/9071896_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart