CH4424852284 ChemBridge Corporation

[2-(2,4-dimethylphenoxy)ethyl]ethylamine hydrochloride

  • MDL number MFCD13186642
  • Cas Number : 1349717-40-3
  • Molecular Weight : 229.7463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071911

Safety Information

MSDS https://www.hit2lead.com/sds/9071911_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart