CH4424852289 ChemBridge Corporation

2-[3-(1H-imidazol-1-yl)propoxy]-3-methoxybenzaldehyde hydrochloride

  • MDL number MFCD13186643
  • Cas Number : 1349709-05-2
  • Molecular Weight : 296.7494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071938

Safety Information

MSDS https://www.hit2lead.com/sds/9071938_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart