CH4424852301 ChemBridge Corporation

N-(3-chlorobenzyl)-2-methyl-2-propanamine hydrochloride

  • MDL number MFCD07109886
  • Cas Number : 1050076-08-8
  • Molecular Weight : 234.1654

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9072027

Safety Information

MSDS https://www.hit2lead.com/sds/9072027_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart