CH4428741591 ChemBridge Corporation

tert-butyl 2-(2-aminoethyl)-1-piperidinecarboxylate hydrochloride

  • MDL number MFCD28122921
  • Cas Number : 1638221-23-4
  • Molecular Weight : 264.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4004161

Safety Information

MSDS https://www.hit2lead.com/sds/4004161_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart