CH4428741602 ChemBridge Corporation

methyl 3-bromo-1H-1,2,4-triazole-5-carboxylate hydrobromide

  • MDL number MFCD28122928
  • Cas Number : 1638221-30-3
  • Molecular Weight : 286.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HBr
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4091215

Safety Information

MSDS https://www.hit2lead.com/sds/4091215_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart