CH4428741614 ChemBridge Corporation

[(2,2-dimethyl-2,3-dihydro-1-benzofuran-5-yl)methyl]amine hydrochloride

  • MDL number MFCD09864192
  • Cas Number : 903556-73-0
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4000011

Safety Information

MSDS https://www.hit2lead.com/sds/4000011_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart