CH4428741623 ChemBridge Corporation

2-(chloromethyl)-1,3-benzothiazole hydrochloride

  • MDL number MFCD13186104
  • Cas Number : 1215284-39-1
  • Molecular Weight : 220.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4016924

Safety Information

MSDS https://www.hit2lead.com/sds/4016924_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart