CH4428741649 ChemBridge Corporation

[(6-chloro[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl]amine hydrochloride

  • MDL number MFCD19103499
  • Cas Number : 518050-85-6
  • Molecular Weight : 220.0593

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043237

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4043237
Availability details for
Quantity
  • Availability
  • Add to Cart