CH4428741658 ChemBridge Corporation

4-(trifluoromethoxy)piperidine hydrochloride

  • MDL number MFCD23382079
  • Cas Number : 1612172-50-5
  • Molecular Weight : 205.6058

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4056201

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4056201
Availability details for
Quantity
  • Availability
  • Add to Cart