CH4428741743 ChemBridge Corporation

2-hydrazino-1-(3-methylbutyl)-1H-benzimidazole dihydrochloride

  • MDL number MFCD08163661
  • Cas Number : 1215598-66-5
  • Molecular Weight : 291.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4091212

Safety Information

MSDS https://www.hit2lead.com/sds/4091212_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart