CH4428741752 ChemBridge Corporation

(5-bromo-2-methoxybenzyl)methylamine hydrobromide

  • MDL number MFCD05982323
  • Cas Number : 1049789-48-1
  • Molecular Weight : 311.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4091221

Safety Information

MSDS https://www.hit2lead.com/sds/4091221_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart