CH4428741756 ChemBridge Corporation

1-isopropyl-1,4-diazepane dihydrochloride

  • MDL number MFCD11841233
  • Cas Number : 851048-47-0
  • Molecular Weight : 215.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4091225

Safety Information

MSDS https://www.hit2lead.com/sds/4091225_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart