CH4432438568 ChemBridge Corporation

1-(aminomethyl)-4,4-difluorocyclohexanol hydrochloride

  • MDL number MFCD28168089
  • Cas Number : 1227808-08-3
  • Molecular Weight : 201.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038502

Safety Information

MSDS https://www.hit2lead.com/sds/4038502_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart