CH4432438573 ChemBridge Corporation

4-bromo-5-(2-thienyl)-1H-pyrazole

  • MDL number MFCD26647573
  • Molecular Weight : 229.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035003

Safety Information

MSDS https://www.hit2lead.com/sds/4035003_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart