CH4432438580 ChemBridge Corporation

(3-exo)-8-azabicyclo[3.2.1]octane-3-carboxylic acid hydrochloride

  • MDL number MFCD27956881
  • Cas Number : 1523530-50-8
  • Molecular Weight : 191.6553

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4054340

Safety Information

MSDS https://www.hit2lead.com/sds/4054340_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart