CH4445001645 ChemBridge Corporation

isothiazole

  • MDL number MFCD00020818
  • Cas Number : 288-16-4
  • Molecular Weight : 85.1276

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036435

Safety Information

MSDS https://www.hit2lead.com/sds/4036435_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart