CH4454669593 ChemBridge Corporation

4,5,6,7-tetrahydroisoxazolo[4,3-c]pyridine hydrochloride

  • MDL number MFCD28895348
  • Cas Number : 1955493-65-8
  • Molecular Weight : 160.6015

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4047970

Safety Information

MSDS https://www.hit2lead.com/sds/4047970_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart