CH4458379377 ChemBridge Corporation

methyl rac-(1R,5S,6r)-3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride

  • MDL number MFCD11099878
  • Cas Number : 1212063-26-7
  • Molecular Weight : 177.6287

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042300

Safety Information

MSDS https://www.hit2lead.com/sds/4042300_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart