CH4458379391 ChemBridge Corporation

[2-(3,4-dichlorophenoxy)ethyl]amine hydrochloride

  • MDL number MFCD18205885
  • Cas Number : 39959-91-6
  • Molecular Weight : 242.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3005342

Safety Information

MSDS https://www.hit2lead.com/sds/3005342_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart