CH4458379398 ChemBridge Corporation

tert-butyl trans-(2-hydroxymethyl)cyclopropylcarbamate

  • MDL number MFCD03844612
  • Cas Number : 177472-54-7
  • Molecular Weight : 187.2362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4046832

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4046832
Availability details for
Quantity
  • Availability
  • Add to Cart