CH4491478830 ChemBridge Corporation

3H-imidazo[4,5-c]pyridin-7-amine

  • MDL number MFCD13175920
  • Cas Number : 792138-97-7
  • Molecular Weight : 134.1386

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4048232

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4048232
Availability details for
Quantity
  • Availability
  • Add to Cart