CH4509506644 ChemBridge Corporation

rac-(1S,2R,4R,6R)-6-hydroxy-7-oxabicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD29924203
  • Cas Number : 2008714-25-6
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4108218

Safety Information

MSDS https://www.hit2lead.com/sds/4108218_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart