CH4509506663 ChemBridge Corporation

4-methyl-1,4-diazepan-5-one hydrochloride

  • MDL number MFCD16872041
  • Cas Number : 329794-41-4
  • Molecular Weight : 164.6332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036421

Safety Information

MSDS https://www.hit2lead.com/sds/4036421_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart