CH4514701318 ChemBridge Corporation

4-bromo-2-naphthol

  • MDL number MFCD01712260
  • Cas Number : 5498-31-7
  • Molecular Weight : 223.066

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038940

Safety Information

MSDS https://www.hit2lead.com/sds/4038940_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart